Learn More
Recombinant VHH Alpaca, anti-Mouse, Unconjugated, Clone: CTK0105, Proteintech ChromoTek
Alpaca Monoclonal Antibody
Brand:  Chromotek SMSG2BCYS2-1-500
Product Code. 17333493
  • 625.25 EUR / 500µL
Description
500 μg Nano-Secondary™ for mouse IgG2b: alpaca monoclonal Nanobody, Fc-specific VHH, for 2x Cys conjugation, clone [CTK0105].
Specifications
Specifications
Recombinant VHH
Monoclonal
Unconjugated
500 μg
Mouse
Immunofluorescence, Western Blot, Flow Cytometry
CTK0105
Alpaca
Secondary