Learn More
enQuireBio™ Recombinant Human Steroid 17-alpha-hydroxylase / 17,20 lyase Protein
Product Code. 15938569
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
Product Code. 15938569
Brand: enQuireBio™ QP5905ec100ug
A DNA sequence encoding the Homo sapiens (Human) Steroid 17-alpha-hydroxylase/17,20 lyase, was expressed in the hosts and tags indicated.
Specifications
Accession Number NP_000093.1
Gene ID (Entrez) 1586
Name Steroid 17-alpha-hydroxylase / 17,20 lyase Protein
Quantity 100 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol CYP17A1
Protein Length Partial (see sequence information for more details).
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His
Sequence GAKYPKSLLS LPLVGSLPFL PRHGHMHNNF FKLQKKYGPI YSVRMGTKTT VIVGHHQLAK EVLIKKGKDF SGRPQMATLD IASNNRKGIA FADSGAHWQL HRRLAMATFA LFKDGDQKLE KIICQEISTL CDMLATHNGQ SIDISFPVFV AVTNVISLIC FNTSYKNGDP ELNVIQNYNE GIIDNLSKDS LVDLVPWLKI FPNKTLEKLK SHVKIRNDLL NKILENYKEK FRSDSITNML DTLMQAKMNS DNGNAGPDQD SELLSDNHIL TTIGDIFGAG VETTTSVVKW TLAFLLHNPQ VKKKLYEEID QNVGFSRTPT ISDRNRLLLL EATIREVLRL RPVAPMLIPH KANVDSSIGE FAVDKGTEVI INLWALHHNE KEWHQPDQFM PERFLNPAGT QLISPSVSYL PFGAGPRSCI GEILARQELF LIMAWLLQRF DLEVPDDGQL PSLEGIPKVV FLIDSFKVKI KVRQAWREAQ AEGST
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 80% as determined by SDS-PAGE.