missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Noggin GMP Protein, CF

Product Code. 32058414
Change view
Click to view available options
Quantity:
1 mg
50 μg
Unit Size:
1mg
50µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32058414 1 mg 1mg
32058404 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 32058414 Supplier R&D Systems™ Supplier No. BTNOGGMP01M
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Recombinant Protein

Measured by its ability to inhibit BMP-4-induced alkaline phosphatase production by ATDC5 mouse chondrogenic cells. The ED50 for this effect is 0.0200-0.240 μg/mL.

TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q13253.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in Sodium Acetate with Trehalose.
Gene ID (Entrez) 9241
Molecular Weight (g/mol) Observed: 27 kDa, under reducing conditions., Theroretical: 23 kDa (monomer)
Quantity 1 mg
Source E. coli-derived human Noggin protein Gln28 - Cys232 with an N-terminal Met Produced using non-animal reagents in an animal-free laboratory. Manufactured and tested under cGMP guidelines.
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. A minimum of 12 months when stored at ≤-20°C as supplied. Refer to lot specific COA for the Use by Date. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, ≤-20°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias NOG, noggin, SYM1, symphalangism 1 (proximal), synostoses (multiple) syndrome 1, SYNS1, SYNS1A
Conjugate Unconjugated
Reconstitution Reconstitute at 500 μg/mL in water.
Product Line R&D Systems
Purity or Quality Grade >97%, by SDS-PAGE with quantitative densitometry by Coomassie™ Blue Staining. The molecular weight by mass spectrometry is 46338 Da ± 5 Da
Protein Noggin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.