missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human MMP-14/MT1-MMP Protein

Product Code. p-200083638 Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
10 μg
100 μg
500 μg
Unit Size:
100µg
10µg
1mg
500µg
This item is not returnable. View return policy

Product Code. 18679088

Brand: Novus Biologicals™ NBP261379100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Test in Process.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) SDS-PAGE
Formulation 0.2 μm filtered solution in 20 mM Tris-HCl, pH 7.4, 300 mM NaCl, 3 mM CaCl2, 10 uM ZnCl2
Gene ID (Entrez) 4323
Molecular Weight (g/mol) MolecularWeight-theroretical: 29.6 kDa
Quantity 100 μg
Storage Requirements Store at -20°C. Avoid freeze/thaw cycles.
Endotoxin Concentration Less than 1 EU/μg of MMP-14/MT1-MMP as determined by LAL method.
Gene Alias EC 3.4.24, EC 3.4.24.80, matrix metallopeptidase 14 (membrane-inserted), matrix metalloproteinase 14 (membrane-inserted), matrix metalloproteinase-14, membrane type 1 metalloprotease, Membrane-type matrix metalloproteinase 1, Membrane-type-1 matrix metalloproteinase, MMP-14, MMP- x 1, MT1MMP, MT1-MMPMTMMP1, MT-MMP 1
Gene Symbol MMP14
Research Category Cancer, Extracellular Matrix
Purity or Quality Grade >95%, by SDS-PAGE and HPLC
Protein MMP-14/MT1-MMP
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.