missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL-22R / IL22RA1 Protein
A DNA sequence encoding the Homo sapiens (Human) IL-22R / IL22RA1, was expressed in the hosts and tags indicated.
160.00 EUR - 2090.00 EUR
Specifications
Accession Number | NP_067081.2 |
---|---|
Gene ID (Entrez) | 58985 |
Name | IL-22R / IL22RA1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16041181
|
enQuireBio™
QP7852-EC-100UG |
100 μg |
463.00 EUR
100µg |
Estimated Shipment: 01-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16061181
|
enQuireBio™
QP7852-EC-1MG |
1 mg |
2090.00 EUR
1mg |
Estimated Shipment: 01-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16051181
|
enQuireBio™
QP7852-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16071181
|
enQuireBio™
QP7852-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16081181
|
enQuireBio™
QP7852-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16091181
|
enQuireBio™
QP7852-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_067081.2 | |
IL-22R / IL22RA1 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
58985 | |
Research Use Only | |
IL22RA1 | |
Recombinant Protein | |
E. coli | |
SYRYVTKPPA PPNSLNVQRV LTFQPLRFIQ EHVLIPVFDL SGPSSLAQPV QYSQIRVSGP REPAGAPQRH SLSEITYLGQ PDISILQPSN VPPPQILSPL SYAPNAAPEV GPPSYAPQVT PEAQFPFYAP QAISKVQPSS YAPQATPDSW PPSYGVCMEG SGKDSPTGTL SSPKHLRPKG QLQKEPPAGS CMLGGLSLQE VTSLAMEESQ EAKSLHQPLG ICTDRTSDPN VLHSGEEGTP QYLKGQLPLL SSVQIEGHPM SLPLQPPSRP CSPSDQGPSP WGLLESLVCP KDEAKSPAPE TSDLEQPTEL DSLFRGLALT VQWE | |
Greater than 90% as determined by SDS-PAGE. |