missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL-22R / IL22RA1 Protein
A DNA sequence encoding the Homo sapiens (Human) IL-22R / IL22RA1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7852-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_067081.2 | |
IL-22R / IL22RA1 Protein | |
Research Use Only | |
IL22RA1 | |
Recombinant Protein | |
E. coli | |
SYRYVTKPPA PPNSLNVQRV LTFQPLRFIQ EHVLIPVFDL SGPSSLAQPV QYSQIRVSGP REPAGAPQRH SLSEITYLGQ PDISILQPSN VPPPQILSPL SYAPNAAPEV GPPSYAPQVT PEAQFPFYAP QAISKVQPSS YAPQATPDSW PPSYGVCMEG SGKDSPTGTL SSPKHLRPKG QLQKEPPAGS CMLGGLSLQE VTSLAMEESQ EAKSLHQPLG ICTDRTSDPN VLHSGEEGTP QYLKGQLPLL SSVQIEGHPM SLPLQPPSRP CSPSDQGPSP WGLLESLVCP KDEAKSPAPE TSDLEQPTEL DSLFRGLALT VQWE | |
Greater than 90% as determined by SDS-PAGE. |
58985 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |