missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IL-22R / IL22RA1 Protein

A DNA sequence encoding the Homo sapiens (Human) IL-22R / IL22RA1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7852-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16061181

  • 2090.00 EUR / 1mg
Estimated Shipment: 10-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_067081.2
IL-22R / IL22RA1 Protein
Research Use Only
IL22RA1
Recombinant Protein
E. coli
SYRYVTKPPA PPNSLNVQRV LTFQPLRFIQ EHVLIPVFDL SGPSSLAQPV QYSQIRVSGP REPAGAPQRH SLSEITYLGQ PDISILQPSN VPPPQILSPL SYAPNAAPEV GPPSYAPQVT PEAQFPFYAP QAISKVQPSS YAPQATPDSW PPSYGVCMEG SGKDSPTGTL SSPKHLRPKG QLQKEPPAGS CMLGGLSLQE VTSLAMEESQ EAKSLHQPLG ICTDRTSDPN VLHSGEEGTP QYLKGQLPLL SSVQIEGHPM SLPLQPPSRP CSPSDQGPSP WGLLESLVCP KDEAKSPAPE TSDLEQPTEL DSLFRGLALT VQWE
Greater than 90% as determined by SDS-PAGE.
58985
1 mg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers