missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Glutamate carboxypeptidase 2 Protein
A DNA sequence encoding the Homo sapiens (Human) Glutamate carboxypeptidase 2, was expressed in the hosts and tags indicated.
160.00 EUR - 2090.00 EUR
Specifications
Accession Number | NP_001014986.1 |
---|---|
Gene ID (Entrez) | 2346 |
Name | Glutamate carboxypeptidase 2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16066151
|
enQuireBio™
QP8726-EC-100UG |
100 μg |
463.00 EUR
100µg |
Estimated Shipment: 08-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16086151
|
enQuireBio™
QP8726-EC-1MG |
1 mg |
2090.00 EUR
1mg |
Estimated Shipment: 08-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16076151
|
enQuireBio™
QP8726-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16096151
|
enQuireBio™
QP8726-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16006161
|
enQuireBio™
QP8726-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16016161
|
enQuireBio™
QP8726-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001014986.1 | |
Glutamate carboxypeptidase 2 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
2346 | |
Research Use Only | |
FOLH | |
Recombinant Protein | |
E. coli | |
EATNITPKHN MKAFLDELKA ENIKKFLYNF TQIPHLAGTE QNFQLAKQIQ SQWKEFGLDS VELAHYDVLL SYPNKTHPNY ISIINEDGNE IFNTSLFEPP PPGYENVSDI VPPFSAFSPQ GMPEGDLVYV NYARTEDFFK LERDMKINCS GKIVIARYGK VFRGNKVKNA QLAGAKGVIL YSDPADYFAP GVKSYPDGWN LPGGGVQRGN ILNLNGAGDP LTPGYPANEY AYRRGIAEAV GLPSIPVHPI GYYDAQKLLE KMGGSAPPDS SWRGSLKVPY NVGPGFTGNF STQKVKMHIH STNEVTRIYN VIGTLRGAVE PDRYVILGGH RDSWVFGGID PQSGAAVVHE IVRSFGTLKK EGWRPRRTIL FASWDAEEFG LLGSTEWAEE NSRLLQERGV AYINADSSIE GNYTLRVDCT PLMYSLVHNL TKELKSPDEG FEGKSLYESW TKKSPSPEFS GMPRISKLGS GNDFEVFFQR LGIASGRARY TKNWETNKFS GYPLYHSVYE TYELVEKFYD PMFKYHLTVA QVRGGMVFEL ANSIVLPFDC RDYAVVLRKY ADKIYSISMK HPQEMKTYSV SFDSLFSAVK NFTEIASKFS ERLQDFDKSN PIVLRMMNDQ LMFLERAFID PLGLPDRPFY RHVIYAPSSH NKYAGESFPG IYDALFDIES KVDPSKAWGE VKRQIYVAAF TVQAAAETLS EVA | |
Greater than 90% as determined by SDS-PAGE. |