missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Glutamate carboxypeptidase 2 Protein
A DNA sequence encoding the Homo sapiens (Human) Glutamate carboxypeptidase 2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8726-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001014986.1 | |
Glutamate carboxypeptidase 2 Protein | |
Research Use Only | |
FOLH | |
Recombinant Protein | |
E. coli | |
EATNITPKHN MKAFLDELKA ENIKKFLYNF TQIPHLAGTE QNFQLAKQIQ SQWKEFGLDS VELAHYDVLL SYPNKTHPNY ISIINEDGNE IFNTSLFEPP PPGYENVSDI VPPFSAFSPQ GMPEGDLVYV NYARTEDFFK LERDMKINCS GKIVIARYGK VFRGNKVKNA QLAGAKGVIL YSDPADYFAP GVKSYPDGWN LPGGGVQRGN ILNLNGAGDP LTPGYPANEY AYRRGIAEAV GLPSIPVHPI GYYDAQKLLE KMGGSAPPDS SWRGSLKVPY NVGPGFTGNF STQKVKMHIH STNEVTRIYN VIGTLRGAVE PDRYVILGGH RDSWVFGGID PQSGAAVVHE IVRSFGTLKK EGWRPRRTIL FASWDAEEFG LLGSTEWAEE NSRLLQERGV AYINADSSIE GNYTLRVDCT PLMYSLVHNL TKELKSPDEG FEGKSLYESW TKKSPSPEFS GMPRISKLGS GNDFEVFFQR LGIASGRARY TKNWETNKFS GYPLYHSVYE TYELVEKFYD PMFKYHLTVA QVRGGMVFEL ANSIVLPFDC RDYAVVLRKY ADKIYSISMK HPQEMKTYSV SFDSLFSAVK NFTEIASKFS ERLQDFDKSN PIVLRMMNDQ LMFLERAFID PLGLPDRPFY RHVIYAPSSH NKYAGESFPG IYDALFDIES KVDPSKAWGE VKRQIYVAAF TVQAAAETLS EVA | |
Greater than 90% as determined by SDS-PAGE. |
2346 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |