missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human COP9 signalosome complex subunit 2 Protein

Product Code. 16090641
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16090641 1 mg 1mg
16080641 10 μg 10µg
16020651 50 μg 50µg
16070641 100 μg 100µg
16000651 200 μg 200µg
16010651 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16090641 Supplier enQuireBio™ Supplier No. QP7761ec1mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens (Human) COP9 signalosome complex subunit 2, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_001137359.1
Gene ID (Entrez) 9318
Name COP9 signalosome complex subunit 2 Protein
Quantity 1 mg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol COPS2
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His-SUMO
Sequence MSDMEDDFMC DDEEDYDLEY SEDSNSEPNV DLENQYYNSK ALKEDDPKAA LSSFQKVLEL EGEKGEWGFK ALKQMIKINF KLTNFPEMMN RYKQLLTYIR SAVTRNYSEK SINSILDYIS TSKQMDLLQE FYETTLEALK DAKNDRLWFK TNTKLGKLYL EREEYGKLQK ILRQLHQSCQ TDDGEDDLKK GTQLLEIYAL EIQMYTAQKN NKKLKALYEQ SLHIKSAIPH PLIMGVIREC GGKMHLREGE FEKAHTDFFE AFKNYDESGS PRRTTCLKYL VLANMLMKSG INPFDSQEAK PYKNDPEILA MTNLVSAYQN NDITEFEKIL KTNHSNIMDD PFIREHIEEL LRNIRTQVLI KLIKPYTRIH IPFISKELNI DVADVESLLV QCILDNTIHG RIDQVNQLLE LDHQKRGGAR YTALDKWTNQ LNSLNQAVVS KLA
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.