missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human C1qTNF1 Protein
Description
The Recombinant Human C1qTNF1 Protein is derived from NS0. The Recombinant Human C1qTNF1 Protein has been validated for the following applications: Bioactivity.
Order Info
The product is shipped at ambient temperature.
Specifications
Specifications
| Accession Number | Q9BXJ1 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS and NaCl |
| Gene ID (Entrez) | 114897 |
| Molecular Weight (g/mol) | 30 kDa, reducing conditions |
| Name | C1q and tumor necrosis factor related protein 1, CTRP1, CTRP1G protein-coupled receptor-interacting protein, FLJ90694, G protein coupled receptor interacting protein, GIPcomplement C1q tumor necrosis factor-related protein 1, ZSIG37 |
| Quantity | 50 μg |
| Source | Mouse myeloma cell line,NS0-derived human CTRP1/C1qTNF1 protein Arg26-Pro281,with a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Regulatory Status | RUO |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?