missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Active PKA C alpha Protein
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Description
The activity of PKA C alpha is typically 1626-2201 nmol/min/mg using a synthetic peptide substrate.
Specifications
Specifications
| Components | Supplied in 50mM Tris-HCl (pH 7.5), 150mM NaCl, 0.25mM DTT, 0.1mM EGTA, 0.1mM EDTA, 0.1mM PMSF, and 25% glycerol. |
| Purity | >90%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain (at 5 μg / lane.) |
| For Use With (Application) | Bioactivity |
| Storage Buffer | Supplied in 50 mM Tris-HCl (pH 7.5), 150 mM NaCl, 10 mM Glutathione, 0.25 mM DTT, 0.1 mM EDTA, 0.1 mM PMSF, and 25% Glycerol. |
| Content And Storage | This product is stable at <= -70°C for up to 1 year from the date of receipt. For optimal storage, aliquot into smaller quantities after centrifugation and store (at recommended temperature. Avoid repeated freeze-thaw cycles.) |
| Accession Number | NM_002730 |
| Gene ID (Entrez) | 5566 |
| Gene Symbol | PRKACA |
| Gene Alias | cAMP-dependent protein kinase catalytic subunit alpha, Cs, EC 2.7.11, EC 2.7.11.11, MGC102831, MGC48865, Pk. C-alpha, PKACA, PKCA1, PKCD, PRKACA, protein kinase A catalytic subunit, protein kinase, cAMP-dependent, catalytic, alpha |
| Source | Spodoptera frugiperda, Sf 9 (baculovirus)-derived human Pk. C alpha protein |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction