missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant HIV-1 gag p17-p24 Protein
Specifications
Specifications
| Name | HIV-1 gag p17-p24 Protein |
| Quantity | 100 μg |
| Regulatory Status | RUO |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | HIV |
| Species | E. coli |
| Protein Tag | Untagged |
| Buffer | 20mM PBS pH 7.8, 20mM NaCl 0.5M, 1mM DTT and 8M urea. |
| Purity or Quality Grade | Greater than 95.0% as determined by HPLC analysis and SDS-PAGE. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?