missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli 30S ribosomal protein S11 Protein

Product Code. 16035621
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16035621 1 mg 1mg
16025621 10 μg 10µg
16065621 50 μg 50µg
16015621 100 μg 100µg
16045621 200 μg 200µg
16055621 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16035621 Supplier enQuireBio™ Supplier No. QP8621ec1mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Escherichia coli (strain K12) 30S ribosomal protein S11, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_417756.1
Gene ID (Entrez) 31655987
Name 30S ribosomal protein S11 Protein
Quantity 1 mg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol rpsK
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity E. coli
Species E. coli
Protein Tag His
Sequence AKAPIRARKR VRKQVSDGVA HIHASFNNTI VTITDRQGNA LGWATAGGSG FRGSRKSTPF AAQVAAERCA DAVKEYGIKN LEVMVKGPGP GRESTIRALN AAGFRITNIT DVTPIPHNGC RPPKKRRV
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.