Learn More
R&D Systems™ Recombinant Cynomolgus/Rhesus Macaque Nectin-1 Protein
Product Code. 18768376
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18768376 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18768376 Supplier R&D Systems™ Supplier No. 10287N1100
 Restricted Item
Estimated Shipment: 04-12-2026
Fc Chimera
Measured by its binding ability in a functional ELISA. When Recombinant Human Nectin-3 His-tag (Catalog # 3064-N3 ) is immobilized at 1 μg/mL (100 μL/well), Recombinant Cynomolgus Monkey/Rhesus Macaque Nectin-1 Fc Chimera (Catalog # 10287-N1) binds with an ED50 of 2.5-20 ng/mL.
TRUSTED_SUSTAINABILITY
Specifications
Accession Number XP_005579992.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 5818
Molecular Weight (g/mol) M.W. (Observed): 87-101 kDa, reducing conditions; M.W. (theoretical): 62 kDa
Name CD111, CD111 antigen, CLPED1ectodermal dysplasia 4 (Margarita Island type), ED4, Herpes virus entry mediator C, Herpesvirus entry mediator C, Herpesvirus Ig-like receptor, HIgRPVRR, HveC, HVECpoliovirus receptor-like 1, MGC142031, Nectin1, Nectin-1, OFC7nectin 1, poliovirus receptor-related 1 (herpesvirus entry mediator C), poliovirus receptor-related protein 1, PRR, PRR1, PRR1MGC16207, PVRL1, PVRR1, PVRR1nectin-1, SK-12
Quantity 100 μg
Source Chinese Hamster Ovary cell line, CHO-derived Nectin-1 protein Cynomolgus Monkey/Rhesus Macaque Nectin-1 (Gln31-Gly349) (N-terminus) Accession # XP_005579992.1 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Conjugate Unconjugated
Species Cynomolgus Monkey, Rhesus macaque
Reconstitution Reconstitute at 1 mg/mL in PBS.
Buffer Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Endotoxin Level <0.10 EU per 1 μg of the protein by the LAL method.
Product Line R&D Systems
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein Nectin-1/PVRL1