missing translation for 'onlineSavingsMsg'
Learn More

asecos™ Recirculating Air Safety Storage Cabinet

Product Code. 16238370 Shop All asecos Products
Change view
Click to view available options
Weight (English):
433.43 lb.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Weight (English) Unit Size
16238370 433.43 lb. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16238370 Supplier asecos™ Supplier No. 3293900130614
  • ONLINE EXCLUSIVE PRICE
    6275.00 EUR
    5386.00 EUR / Pack of 1
    Save 889.00 EUR
    14% Off
 Restricted Item
Estimated Shipment: 02-03-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Safe and approved storage with powder-coated sheet steel for aggressive, non flammable hazardous materials in working areas

Cabinet body

  • Robust construction and longevity: cabinet body made of powder-coated sheet steel, safety elements assembled outside the storage compartment for increased protection against corrosion
  • Two in one: two hermetically separated storage compartments allow the isolated storage of acids and bases in one cabinet
  • No unauthorised use: doors lockable with profile cylinder (integration in an existing locking system possible)
  • Easy alignment: adjusting aids to compensate for uneven floor

Recirculating air filter system

  • Ideal for the direct installation in working areas – very low noise, only approx. 39 dB (A)
  • With multi-stage wideband filter
  • Monitoring electronics for extraction incl. cool-device cable and power plug
  • Optical and acoustical alarm including potential-free alarm contact
  • Display with innovative touch panel
  • Manual check of the filter saturation

Specifications

Certifications/Compliance DIN EN 16121/16122 Conformity, GS Certified, CE Compliant
Configuration 4 x Pull-out Shelf with Plastic Sump
Depth (English) Interior 22.24 in.
Depth (Metric) Exterior 615 mm
Depth (Metric) Interior 565 mm
Dimensions (D x W x H) Interior 565 x 485 x 1865 mm
Dimensions (D x W x H) 615 x 600 x 2300 mm
Door Style Wing
For Use With (Application) Storage of Aggressive, Non flammable Hazardous Materials
Height (English) Interior 73.43 in.
Height (Metric) Exterior 2300 mm
Height (Metric) Interior 1865 mm
Includes Interior Fittings
Model SLX.230.060.MH
No. of Doors 2
No. of Shelves 4
Shelf Type Pull-out Shelf
Surface Coating Melamin Resin Laminated
Weight (English) 433.43 lb.
Weight (Metric) 196.60 kg
Width (English) Interior 19.09 in.
Width (Metric) Exterior 600 mm
Width (Metric) Interior 485 mm
Color Light Gray
Depth (English) Exterior 24.21 in.
Height (English) Exterior 90.55 in.
Height (Metric) Opening 1865 mm
Material Chipboard
Product Type Recirculating Air Safety Storage Cabinet
Width (English) Exterior 23.62 in.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.