Learn More
Thermo Electron LED GmbH Reagent Reservoir
Description
The reagent reservoir from Thermo Electron LED GmbH is made of polypropylene and has eight wells.
- Autoclavable
- Provide variety of solutions to suit all pipetting procedures
- Reagent reservoirs designed to provide solutions tailored to procedures
Specifications
Specifications
| Color | White |
| Capacity (Metric) | 60 mL |
| Material | Polypropylene |
| Sterility | Non-sterile |
| Type | Reagent Reservoir |
| Quantity | 10 Pk. |
| Autoclavable | Autoclavable |
| Channels | 8 |
| Disposable | Reusable |
| Bottom Shape | Flat |