missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Reagent A Fehling Solution, Chem Lab™
Specifications
Specifications
| Color | Blue |
| Boiling Point | 100°C |
| Melting Point | 0°C |
| Composition | 69.2 g CuSO4.5H2O + 1mL H2SO4 95-97% / l H2O |
| Density | 1.01g/mL |
| Solubility Information | Water Soluble |
| Odor | Odorless |
| pH | 4 to 5 |
| Quantity | 1 L |
| Physical Form | Liquid |
Safety and Handling
Product Identifier
- Fehling A reagent
- Warning
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H410-Very toxic to aquatic life with long lasting effects.
- P273-Avoid release to the environment.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contain: copper sulphate
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?