Learn More
Thermo Scientific™ Reacti-Vap™ Evaporator PTFE-Coated Needles
Product Code. 11507490 Shop All Thermo Scientific Products
Change view
Click to view available options
Length (Metric) Needle:
102 mm
152 mm
Unit Size:
Pack of 9
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Needle Unit Size
11507490 102 mm Pack of 9
11517490 152 mm Pack of 9
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11507490 Supplier Thermo Scientific™ Supplier No. TS18784
Estimated Shipment: 27-11-2026
Use these needles to deliver gas and evaporate samples on the Thermo Scientific™ Reacti-Vap™ Evaporator.
Use Thermo Scientific™ Reacti-Vap™ PTFE-Coated Needles to deliver gas and evaporate samples on Reacti-Vap Evaporators. These blunt-ended, 19-gauge, stainless steel needles reduce cross-contamination and corrosion when evaporating solvents that contain strong acids. The needles were specifically designed for Reacti-Vap Evaporators.
TRUSTED_SUSTAINABILITY
Specifications
Material PTFE-coated
Description 9 Pack
Needle Gauge 19 G
Length (English) Needle 4 in.
Product Type Syringe Needle
Length (Metric) Needle 102 mm
For Use With (Equipment) Reacti-Vap Evaporators