Learn More
Thermo Scientific™ Reacti-Therm™ Magnetic Stirrers for Reacti-Vial™ Small Reaction Vials
Product Code. 11597450 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
0.3 and 1mL Reacti-Vials
3, 5, and 10 mL Reacti-Vials
Description:
6 Pack
Unit Size:
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Description Unit Size
11597450 0.3 and 1mL Reacti-Vials 6 Pack Pack of 6
11587450 3, 5, and 10 mL Reacti-Vials 6 Pack Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11597450 Supplier Thermo Scientific™ Supplier No. TS16010
 Restricted Item
Estimated Shipment: 28-01-2027
Use these triangular stir bars for faster reaction times and smooth mixing in vials of the Thermo Scientific™ Reacti-Therm™ Derivatization System.
Use these triangular stir bars for faster reaction times and smooth mixing of small samples in conical-bottom vials of the Thermo Scientific™ Reacti-Therm™ Derivatization System. The stir bars provide simultaneous stirring of samples in multiple vials when used with a Reacti-Therm Heating/Stirring Module equipped with appropriate Reacti-Block Aluminum Blocks.
TRUSTED_SUSTAINABILITY
Specifications
Description 6 Pack
For Use With (Equipment) 0.3 and 1mL Reacti-Vials