missing translation for 'onlineSavingsMsg'
Learn More

Rb1 Mouse, Clone: 1F8, enQuireBio™

Product Code. p-7156189
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µg
20µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15930778 20 μg 20µg
15940778 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15930778 Supplier EnquireBio Supplier No. 5925MSM1P0

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Specifications

Antigen Rb1
Applications Immunohistochemistry
Cellular Localization Nuclear
Classification Monoclonal
Clone 1F8
Concentration 200μg/mL
Conjugate Unconjugated
Formulation 10mM PBS with 0.05% BSA and 0.05% Azide
Gene Accession No. NP_000312
Gene Symbols RB1
Host Species Mouse
Immunogen Recombinant human Rb1 protein fragment of 283 amino acid residues
Molecular Weight of Antigen 105kDa
Positive Control HeLa, A431 or BT-20 cells. Breast or Colon carcinoma.
Purification Method Protein A/G Purified
Quantity 20 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5925
Content And Storage Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.