missing translation for 'onlineSavingsMsg'
Learn More

Rat IgG2a Kappa Light Chain, Clone: R2a, Isotype control, Novus Biologicals™

Product Code. 18235373 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
0.5 mg
Unit Size:
0.05mg
0.50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18235373 0.05 mg 0.05mg
18290975 0.5 mg 0.50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18235373 Supplier Novus Biologicals Supplier No. NBP1433210.05MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 3 publications

IgG2a Kappa Light Chain Monoclonal specifically detects IgG2a Kappa Light Chain in Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Control.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG2a Kappa Light Chain
Applications Flow Cytometry
Classification Monoclonal
Clone R2a
Conjugate Unconjugated
Dilution Flow Cytometry 1:10 - 1:1000
Formulation Aqueous buffer, may contain carrier protein/stabilizer (proprietary). with 0.09% Sodium Azide
Host Species Rat
Quantity 0.05 mg
Primary or Secondary Primary
Test Specificity The monoclonal rat IgG2a, kappa immunoglobulin is useful as an isotype control for rat IgG2a antibodies.
Target Species Rat
Content And Storage Store at 4C in the dark.
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.