missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Rat B7-2/CD86 (NP_064466) VersaClone cDNA

Product Code. 18749777
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
This item is not returnable. View return policy

Product Code. 18749777

Brand: R&D Systems™ RDC1770

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

B-lymphocyte activation antigen B7-2

The Rat B7-2/CD86 (NP_064466) VersaClone cDNA clone contains the complete ORF for B7-2/CD86, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The B7-2/CD86 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.

Order Info

Ships at ambient temperature

TRUSTED_SUSTAINABILITY

Specifications

Product Type cDNA
Gene B7-2/CD86
Accession Number NP_064466
Concentration 10 μg at 0.2 μg/μL
Formulation cDNA is provided in 10mM Tris-Cl, pH 8.5
Insert Type 955bp
Gene ID (Entrez) 942
Gene Alias Activation B7-2 antigen, B70, B72, B7-2, B7-2 antigen, B-lymphocyte activation antigen B7-2, BU63, CD28 antigen ligand 2, CD28LG2B7-2 antigen), CD86, CD86 antigen, CD86 molecule, CTLA-4 counter-receptor B7.2, FUN-1, LAB72, MGC34413, T-lymphocyte activation antigen CD86
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Rat
Quantity 10 μg
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.