missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Rat alpha 2-Macroglobulin ELISA Kit (Colorimetric)

Product Code. 18638630 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18638630 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18638630 Supplier Novus Biologicals™ Supplier No. NBP260637

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Kit (Colorimetric)

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 0.313 to 20 ng/mL (example only, lot dependent)
Assay Sensitivity 0.1 ng/mL (example only; lot dependent)
Product Type ELISA Kit (Colorimetric)
Sample Type Serum, Plasma, Cell Culture, Urine
For Use With (Application) ELISA, Sandwich ELISA
Gene ID (Entrez) 2
Gene Symbol A2M
Quantity 1 Kit
Research Discipline Neuroscience
Storage Requirements Storage is content dependent.
Synonym alpha-2-M, alpha-2-macroglobulin, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 5, CPAMD5DKFZp779B086, FWP007, S863-7
Target Rat
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.