Learn More
Lonza™ Rapid Endotest - Five Research Samples
Product Code. 11545789
Change view
Click to view available options
Product Type:
Endotoxin Testing Service
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
11545789 Endotoxin Testing Service Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11545789 Supplier Lonza™ Supplier No. BE95103
  • ONLINE EXCLUSIVE PRICE
    1335.00 EUR
    1128.58 EUR / Pack of 5
    Save 206.42 EUR
    15% Off
 Restricted Item
Estimated Shipment: 02-12-2026
Endotoxin testing service that offers a mail-in, pre-paid services for users not requiring testing according to pharmaceutical or medical device regulations.
  • Endotoxin testing service for up to five research samples.
  • Samples are tested at 1:10, 1:100 and 1:1000 dilutions with a positive product control.
  • Mail-in, pre-paid service for users not requiring testing according to pharmaceutical or medical device regulations.
  • Customers can purchase Rapid Endotest packs from Lonza to submit samples to our testing service for basic determination of the endotoxin content in their sample(s).
  • Preliminary screening or inhibition/enhancement tests are not performed on samples submitted through the Pre-paid Service.
  • Unless otherwise noted, samples will be tested using the Kinetic-QCL™ LAL Assay.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Not according to pharmaceutical or medical device regulations.
Description Mail-in, pre-paid testing service.
Product Type Endotoxin Testing Service
For Use With (Application) Endotoxin testing