Learn More
Ramipril, TRC
Specifications
Specifications
| Purity | >95% (HPLC) |
| CAS | 87333-19-5 |
| Molecular Weight (g/mol) | 416.51 |
| Quantity | 100 mg |
| Shipping Condition | Room Temperature |
| Synonym | Ramipril∣(2S,3aS,6aS)-1-[(2S)-2-[[(1S)-1-(Ethoxycarbonyl)-3-phenylpropyl]amino]propanoyl]octahydrocyclopenta[b]pyrrole-2-carboxylic acid∣(2S,3aS,6aS)-1-[(2S)-2-[[(1S)-1-(Ethoxycarbonyl)-3-phenylpropyl]amino]-1-oxopropyl]octahydrocyclopenta[b]pyrrole-2-carboxylic acid∣ [2S-[1[R*(R*)],2α,3aβ,6aβ]]-1-[2-[[1-(Ethoxycarbonyl)-3-phenylpropyl]amino]-1-oxopropyl]octahydrocyclopenta[b]pyrrole-2-carboxylic acid∣Altace∣Cardace∣Delix∣HOE 498∣Hopace∣Pramace∣Quark∣Ramace∣Ramipres∣Ramipril∣Triatec∣Tritace∣Unipril∣Vesdil |