missing translation for 'onlineSavingsMsg'
Learn More

Raloxifene N-Oxide, TRC

Product Code. 30419474
Change view
Click to view available options
Quantity:
5 mg
10 mg
100 mg
Unit Size:
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30419474 10 mg -
30410179 5 mg 5mg
30416295 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30419474 Supplier Toronto Research Chemicals Supplier No. TRCR10005010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 195454-31-0
Molecular Formula C28 H27 N O5 S
Molecular Weight (g/mol) 489.58
Synonym [6-Hydroxy-2-(4-hydroxyphenyl)-1-benzothiophen-3-yl][4-[2-(piperidin-1-yl)ethoxy]phenyl]methanone N-Oxide, Raloxifene Hydrochloride Imp. C (EP), Raloxifene N-Oxide
IUPAC Name [6-hydroxy-2-(4-hydroxyphenyl)-1-benzothiophen-3-yl]-[4-[2-(1-oxidopiperidin-1-ium-1-yl)ethoxy]phenyl]methanone
SMILES Oc1ccc(cc1)c2sc3cc(O)ccc3c2C(=O)c4ccc(OCC[N+]5([O-])CCCCC5)cc4

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C28H27NO5S/c30-21-8-4-20(5-9-21)28-26(24-13-10-22(31)18-25(24)35-28)27(32)19-6-11-23(12-7-19)34-17-16-29(33)14-2-1-3-15-29/h4-13,18,30-31H,1-3,14-17H2
Formula Weight 489.161
Percent Purity >95%
Chemical Name or Material (6-Hydroxy-2-(4-hydroxyphenyl)-1-benzothiophen-3-yl)[4-[2-(piperidin-1-yl)ethoxy]phenyl]methanone N-Oxide (Raloxifene N-Oxide)
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.