CAS RN 195454-31-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Raloxifene N-Oxide, TRC
CAS: 195454-31-0 Molecular Formula: C28 H27 N O5 S Molecular Weight (g/mol): 489.58 Synonym: [6-Hydroxy-2-(4-hydroxyphenyl)-1-benzothiophen-3-yl][4-[2-(piperidin-1-yl)ethoxy]phenyl]methanone N-Oxide,Raloxifene Hydrochloride Imp. C (EP),Raloxifene N-Oxide IUPAC Name: [6-hydroxy-2-(4-hydroxyphenyl)-1-benzothiophen-3-yl]-[4-[2-(1-oxidopiperidin-1-ium-1-yl)ethoxy]phenyl]methanone SMILES: Oc1ccc(cc1)c2sc3cc(O)ccc3c2C(=O)c4ccc(OCC[N+]5([O-])CCCCC5)cc4
2
[6-Hydroxy-2-(4-hydroxyphenyl)-1-benzothiophen-3-yl][4-[2-(piperidin-1-yl)ethoxy]phenyl]methanone N-oxide (Raloxifene N-Oxide), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.