Learn More
Grant Instruments™ Raised Shelf for Unstirred Water Baths
Product Code. 11496477
Change view
Click to view available options
Type:
Shelf for 12 L Tanks
Shelf for 18 L Tanks
Shelf for 26 L Tanks
Shelf for 34 L Tanks
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type unitSize
11496477 Shelf for 12 L Tanks Each
11406487 Shelf for 18 L Tanks Each
11416487 Shelf for 26 L Tanks Each
11426487 Shelf for 34 L Tanks Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11496477 Supplier Grant Instruments™ Supplier No. RS14
Only null left
Add to basket
Add to basket
Accommodates shallow vessels. Grant Instruments™ Raised Shelf for Unstirred Water Baths can be used with a variety of other Grant Heating Circulators and Heated Circulating Baths.
  • Choose from a variety of sizes to fit your tank.
  • For use with either Stainless Steel and Plastic tank models
TRUSTED_SUSTAINABILITY
Specifikationer
For Use With (Equipment) Grant Heated Circulating Baths
Type Shelf for 12 L Tanks
Height (Metric) 40 or 78 mm
Produkttitel
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.