Learn More
Fisherbrand™ Radiation Safety Carboy Covers
Product Code. 12307147 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Height (Metric):
9 cm (internal), 60 cm (external)
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) Unit Size
12307147 9 cm (internal), 60 cm (external) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12307147
Estimated Shipment: 15-02-2027
Add to basket
Add to basket
Idea cover for the protection of carboys and laboratory equipment.
  • For the protection of carboys and laboratory equipment
  • An open base allows the cover to be placed over the container or equipment
  • A hinged lid provides easy access
  • Can be used in conjunction with a laboratory safety tray
Specifications
Dimensions (L x W x H) 59 x 38 x 38cm (internal) and 60 x 40 x 40cm (external)
Height (Metric) 9 cm (internal), 60 cm (external)
Length (Metric) 59cm
Product Type Radiation protection carboy cover