missing translation for 'onlineSavingsMsg'
Learn More

Radafaxine hydrochloride, MedChemExpress

Product Code. p-200099936
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16477408 10 mg 10mg
16439887 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16477408 Supplier Medchem Express Supplier No. HY17590
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Radafaxine hydrochloride (GW-353162A) is a DAT (dopamine transporter) and NET(norepinephrine transporter) transporters inhibitor, and nAChR family modulator.

Radafaxine hydrochloride (GW-353162A) is a DAT (dopamine transporter) and NET(norepinephrine transporter) transporters inhibitor, and nAChR family modulator.

Specifications

Chemical Name or Material Radafaxine hydrochloride
Physical Form Solid
CAS 106083-71-0
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C13H19Cl2NO2
For Use With (Application) Neuroscience-Neuromodulation
Synonym GW-353162A BW-306U
Solubility Information DMSO : ≥ 38 mg/mL (130.05 mM) ∣H2O : 33.33 mg/mL (114.07 mM; Need ultrasonic)
SMILES O[C@]1(C2=CC=CC(Cl)=C2)[C@H](C)NC(C)(C)CO1.[H]Cl
Molecular Weight (g/mol) 292.2
Formula Weight 292.2
Percent Purity 99.88%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.