missing translation for 'onlineSavingsMsg'
Learn More

rac Salsolinol, Hydrobromide, TRC

Product Code. 30427639
Change view
Click to view available options
Quantity:
10 mg
100 mg
1 g
Unit Size:
100mg
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30427639 1 g 1g
30424618 100 mg 100mg
30426287 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30427639 Supplier Toronto Research Chemicals Supplier No. TRCS0999951G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59709-57-8
Molecular Formula C10 H13 N O2 . Br H
Molecular Weight (g/mol) 260.13
Synonym (+/-)-Salsolinol hydrobromide - Bio-X TM
IUPAC Name 1-methyl-1,2,3,4-tetrahydroisoquinoline-6,7-diol;hydrobromide
SMILES Br.CC1NCCc2cc(O)c(O)cc12

Specifications

Quantity 1 g
InChI Formula InChI=1S/C10H13NO2.BrH/c1-6-8-5-10(13)9(12)4-7(8)2-3-11-6;/h4-6,11-13H,2-3H2,1H3;1H
Formula Weight 259.0208
Chemical Name or Material (+/-)-Salsolinol, Hydrobromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.