Learn More
rac-Ruxolitinib-d9 (major), TRC
Specifications
Specifications
| Chemical Name or Material | rac-Ruxolitinib-d9 |
| CAS | 2469553-67-9 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C17 H9 D9 N6 |
| Quantity | 2.5 mg |
| Synonym | β-Cyclopentyl-4-(7H-pyrrolo[2,3-d]pyrimidin-4-yl)-1H-pyrazole-1-propanenitrile-d9, INCB 018424-d9 |
| InChI Formula | InChI=1S/C17H18N6/c18-7-5-15(12-3-1-2-4-12)23-10-13(9-22-23)16-14-6-8-19-17(14)21-11-20-16/h6,8-12,15H,1-5H2,(H,19,20,21)/i1D2,2D2,3D2,4D2,12D |
| SMILES | [2H]C1([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C1([2H])C(CC#N)N(N=C2)C=C2C3=NC=NC4=C3C=CN4 |
| IUPAC Name | 3-(1,2,2,3,3,4,4,5,5-nonadeuteriocyclopentyl)-3-[4-(7H-pyrrolo[2,3-d]pyrimidin-4-yl)pyrazol-1-yl]propanenitrile |
| Molecular Weight (g/mol) | 315.42 |
| Show More |