Learn More
rac O-Desmethyl Venlafaxine Beta-D-Glucuronide, TRC
Product Code. 30349995
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30349995 10 mg 10mg
30354471 5 mg 5mg
30353702 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30349995 Supplier Toronto Research Chemicals Supplier No. TRCD29652510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1021933-98-1
Molecular Formula C22 H33 N O8
Molecular Weight (g/mol) 439.5
IUPAC Name (2S,3S,4S,5R)-6-[4-[2-(dimethylamino)-1-(1-hydroxycyclohexyl)ethyl]phenoxy]-3,4,5-trihydroxyoxane-2-carboxylic acid
SMILES CN(C)CC(c1ccc(OC2O[C@@H]([C@@H](O)[C@H](O)[C@H]2O)C(=O)O)cc1)C3(O)CCCCC3
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C22H33NO8/c1-23(2)12-15(22(29)10-4-3-5-11-22)13-6-8-14(9-7-13)30-21-18(26)16(24)17(25)19(31-21)20(27)28/h6-9,15-19,21,24-26,29H,3-5,10-12H2,1-2H3,(H,27,28)/t15?,16-,17-,18+,19-,21?/m0/s1
Formula Weight 439.2206
Chemical Name or Material O-Desmethyl-(rac-venlafaxine) Glucuronide