CAS RN 1021933-98-1
CAS RN 1021933-98-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
rac O-Desmethyl Venlafaxine Beta-D-Glucuronide, TRC
CAS: 1021933-98-1 Molecular Formula: C22 H33 N O8 Molecular Weight (g/mol): 439.5 IUPAC Name: (2S,3S,4S,5R)-6-[4-[2-(dimethylamino)-1-(1-hydroxycyclohexyl)ethyl]phenoxy]-3,4,5-trihydroxyoxane-2-carboxylic acid SMILES: CN(C)CC(c1ccc(OC2O[C@@H]([C@@H](O)[C@H](O)[C@H]2O)C(=O)O)cc1)C3(O)CCCCC3