Learn More
rac N,O-Didesmethyl Venlafaxine-d3, TRC
Specifications
Specifications
| Chemical Name or Material | rac N,O-Didesmethyl Venlafaxine-d3 |
| CAS | 1189468-67-4 |
| Molecular Formula | C15H20D3NO2 |
| Quantity | 10 mg |
| Synonym | 4-[1-(1-Hydroxycyclohexyl)-2-(methyl-d3-amino)ethyl]phenol |
| InChI Formula | InChI=1S/C15H23NO2/c1-16-11-14(12-5-7-13(17)8-6-12)15(18)9-3-2-4-10-15/h5-8,14,16-18H,2-4,9-11H2,1H3/i1D3 |
| SMILES | OC1(CCCCC1)C(CNC([2H])([2H])[2H])C2=CC=C(O)C=C2 |
| Molecular Weight (g/mol) | 252.37 |