missing translation for 'onlineSavingsMsg'
Learn More

rac-N-[(1R)-2-Hydroxy-3-methyl-1-(2-methylpropyl)-3-buten-1-yl]-carbamic Acid 1,1-Dimethylethyl Ester, TRC

Product Code. 30401624
Change view
Click to view available options
Quantity:
5 mg
25 mg
100 mg
Unit Size:
100mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30401624 25 mg 25mg
30391906 100 mg 100mg
30389018 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30401624 Supplier Toronto Research Chemicals Supplier No. TRCH94969825MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material rac-N-[(1R)-2-Hydroxy-3-methyl-1-(2-methylpropyl)-3-buten-1-yl]-carbamic Acid 1,1-Dimethylethyl Ester
Synonym rac-t-Butyl ((4R)-3-Hydroxy-2,6-dimethylhept-1-en-4-yl)carbamate
Quantity 25 mg
Shipping Condition Room Temperature
Molecular Formula C12 H16 N2 O7
Formula Weight 300.096
Molecular Weight (g/mol) 300.265
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.