Learn More
rac Ketorolac 6-Benzoyl Isomer, TRC
Product Code. 30134531
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134531 1 mg 1mg
30012006 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30134531 Supplier Toronto Research Chemicals Supplier No. TRCK2356201MG
 Restricted Item
Estimated Shipment: 08-10-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1026936-07-1
Molecular Formula C15 H13 N O3
Molecular Weight (g/mol) 255.27
Synonym 6-Benzoyl-2,3-dihydro-1H-pyrrolizine-1-carboxylic Acid, (1RS)-6-Benzoyl-2,3-dihydro-1H-pyrrolizine-1-carboxylic Acid
IUPAC Name 6-benzoyl-2,3-dihydro-1H-pyrrolizine-1-carboxylic acid
SMILES OC(=O)C1CCn2cc(cc12)C(=O)c3ccccc3
Specifications
Quantity 1 mg
InChI Formula InChI=1S/C15H13NO3/c17-14(10-4-2-1-3-5-10)11-8-13-12(15(18)19)6-7-16(13)9-11/h1-5,8-9,12H,6-7H2,(H,18,19)
Formula Weight 255.0895
Chemical Name or Material (1RS)-6-Benzoyl-2,3-dihydro-1H-pyrrolizine-1-carboxylic acid