Learn More
rac FTY720 Phosphate, TRC
Product Code. 30136639
Change view
Click to view available options
Quantity:
1 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136639 25 mg 25mg
30010866 1 mg 1mg
30008852 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30136639 Supplier Toronto Research Chemicals Supplier No. TRCF80501025MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 402615-91-2
Molecular Formula C19 H34 N O5 P
Molecular Weight (g/mol) 387.45
Synonym 1,3-Propanediol, 2-amino-2-[2-(4-octylphenyl)ethyl]-, 1-(dihydrogen phosphate), 1,3-Propanediol, 2-amino-2-[2-(4-octylphenyl)ethyl]-, mono(dihydrogen phosphate) (ester) (9CI), FTY 720 phosphate, FTY 720P, FTY-P, Fingolimod phosphate, Fingolimod-P, rac FTY720 Phosphate, Fingolimod O-phosphate
IUPAC Name [2-amino-2-(hydroxymethyl)-4-(4-octylphenyl)butyl] dihydrogen phosphate
SMILES CCCCCCCCc1ccc(CCC(N)(CO)COP(=O)(O)O)cc1
Specifications
Quantity 25 mg
InChI Formula InChI=1S/C19H34NO5P/c1-2-3-4-5-6-7-8-17-9-11-18(12-10-17)13-14-19(20,15-21)16-25-26(22,23)24/h9-12,21H,2-8,13-16,20H2,1H3,(H2,22,23,24)
Formula Weight 387.2175
Percent Purity >95%
Chemical Name or Material rac FTY720 Phosphate
Grade HPLC