missing translation for 'onlineSavingsMsg'
Learn More

rac Fosfomycin-13C3 Benzylamine Salt, TRC

Product Code. 30006589
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30006589 1 mg 1mg
30005366 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30006589 Supplier Toronto Research Chemicals Supplier No. TRCF7275021MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1216461-18-5
Molecular Formula C7 H9 N . 13C3 H7 O4 P
Molecular Weight (g/mol) 248.19
Synonym (3-Methyloxiranyl)phosphonic-13C3 acid benzylamine salt, P-[3-(Methyl-13C)-2-oxiranyl-1,2-13C2]phosphonic acid benzylamine salt, (1,2-Epoxypropyl-13C3)phosphonic acid benzylamine salt, rac-Fosfomycin-13C3 benzylamine salt
IUPAC Name (3-(1^{13}C)methyl(2,3-^{13}C_{2})oxiran-2-yl)phosphonic acid;phenylmethanamine
SMILES [13CH3][13CH]1O[13CH]1P(=O)(O)O.NCc2ccccc2

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C7H9N.C3H7O4P/c8-6-7-4-2-1-3-5-7;1-2-3(7-2)8(4,5)6/h1-5H,6,8H2;2-3H,1H3,(H2,4,5,6)/i;1+1,2+1,3+1
Formula Weight 248.0918
Percent Purity >95%
Chemical Name or Material rac-Fosfomycin-13C3 Benzylamine Salt
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.