CAS RN 1216461-18-5
CAS RN 1216461-18-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
rac Fosfomycin-13C3 Benzylamine Salt, TRC
CAS: 1216461-18-5 Molecular Formula: C7 H9 N . 13C3 H7 O4 P Molecular Weight (g/mol): 248.19 Synonym: (3-Methyloxiranyl)phosphonic-13C3 acid benzylamine salt,P-[3-(Methyl-13C)-2-oxiranyl-1,2-13C2]phosphonic acid benzylamine salt,(1,2-Epoxypropyl-13C3)phosphonic acid benzylamine salt,rac-Fosfomycin-13C3 benzylamine salt IUPAC Name: (3-(1^{13}C)methyl(2,3-^{13}C_{2})oxiran-2-yl)phosphonic acid;phenylmethanamine SMILES: [13CH3][13CH]1O[13CH]1P(=O)(O)O.NCc2ccccc2