Learn More
rac Fosfomycin-13C3 Benzylamine Salt, TRC
Product Code. 30005366
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30005366 10 mg 10mg
30006589 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30005366 Supplier Toronto Research Chemicals Supplier No. TRCF72750210MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1216461-18-5
Molecular Formula C7 H9 N . 13C3 H7 O4 P
Molecular Weight (g/mol) 248.19
Synonym (3-Methyloxiranyl)phosphonic-13C3 acid benzylamine salt, P-[3-(Methyl-13C)-2-oxiranyl-1,2-13C2]phosphonic acid benzylamine salt, (1,2-Epoxypropyl-13C3)phosphonic acid benzylamine salt, rac-Fosfomycin-13C3 benzylamine salt
IUPAC Name (3-(1^{13}C)methyl(2,3-^{13}C_{2})oxiran-2-yl)phosphonic acid;phenylmethanamine
SMILES [13CH3][13CH]1O[13CH]1P(=O)(O)O.NCc2ccccc2
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C7H9N.C3H7O4P/c8-6-7-4-2-1-3-5-7;1-2-3(7-2)8(4,5)6/h1-5H,6,8H2;2-3H,1H3,(H2,4,5,6)/i;1+1,2+1,3+1
Formula Weight 248.0918
Percent Purity >95%
Chemical Name or Material rac-Fosfomycin-13C3 Benzylamine Salt
Grade HPLC