missing translation for 'onlineSavingsMsg'
Learn More

rac Enterodiol-13C3, TRC

Product Code. 30137711
Change view
Click to view available options
Quantity:
1 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137711 10 mg 10mg
30138606 25 mg 25mg
30010550 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137711 Supplier Toronto Research Chemicals Supplier No. TRCE55894210MG
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 918502-74-6
Molecular Formula C1513C3H22O4
Molecular Weight (g/mol) 305.34
SMILES O[13CH2][13CH](Cc1cccc(O)c1)C([13CH2]O)Cc2cccc(O)c2

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C18H22O4/c19-11-15(7-13-3-1-5-17(21)9-13)16(12-20)8-14-4-2-6-18(22)10-14/h1-6,9-10,15-16,19-22H,7-8,11-12H2/i11+1,12+1,15+1
Chemical Name or Material (+/-) Enterodiol 13C3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.