Learn More
rac-cis Dorzolamide Hydrochloride, TRC
Product Code. 17972215
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17972215 100 mg 100mg
17913735 10 mg 10mg
17941795 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17972215 Supplier Toronto Research Chemicals Supplier No. TRCD535115100MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 120279-37-0
Molecular Formula C10H17ClN2O4S3
Molecular Weight (g/mol) 360.9
Synonym (4R,6S)-rel-4-(Ethylamino)-5,6-dihydro-6-methyl-4H-thieno[2,3-b]thiopyran-2-sulfonamide 7,7-Dioxide Monohydrochloride, cis-4-(Ethylamino)-5,6-dihydro-6-methyl-4H-thieno[2,3-b]thiopyran-2-sulfonamide 7,7-Dioxide Monohydrochloride, Dorzolamide Related Compound B USP, Dorzolamide Impurity B (EP) Hydrochloride
IUPAC Name (4R,6S)-4-(ethylamino)-6-methyl-7,7-dioxo-5,6-dihydro-4H-thieno[2,3-b]thiopyran-2-sulfonamide;hydrochloride
SMILES CCNC1CC(S(=O)(=O)C2=C1C=C(S2)S(=O)(=O)N)C.Cl
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C10H16N2O4S3.ClH/c1-3-12-8-4-6(2)18(13,14)10-7(8)5-9(17-10)19(11,15)16;/h5-6,8,12H,3-4H2,1-2H3,(H2,11,15,16);1H/t6-,8+;/m0./s1
Formula Weight 360.004
Percent Purity >95%
Chemical Name or Material (4R,6S)-4-(Ethylamino)-6-methyl-5,6-dihydro-4H-thieno[2,3-b]thiopyran-2-sulfonamide 7,7-dioxide hydrochloride
Grade HPLC