Learn More
rac cis-9,10-Epoxystearic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | rac cis-9,10-Epoxystearic Acid |
| CAS | 24560-98-3 |
| Purity Grade Notes | GC |
| Molecular Formula | C18 H34 O3 |
| Quantity | 50 mg |
| Synonym | (2R,3S)-rel-3-Octyl-2-oxiraneoctanoic Acid, cis-9,10-Epoxyoctadecanoic Acid, (2R,3S)-rel-3-Octyloxiraneoctanoic Acid, cis-3-Octyloxiraneoctanoic Acid, cis-9,10-Epoxyoctadecanoate, cis-9,10-Epoxyoctadecanoic Acid, cis-9,10-Epoxystearic Acid, cis-DL-9,10-Epoxystearic Acid, cis-Epoxystearic Acid |
| InChI Formula | InChI=1S/C18H34O3/c1-2-3-4-5-7-10-13-16-17(21-16)14-11-8-6-9-12-15-18(19)20/h16-17H,2-15H2,1H3,(H,19,20)/t16-,17+/m0/s1 |
| SMILES | CCCCCCCC[C@@H]1O[C@@H]1CCCCCCCC(=O)O |
| IUPAC Name | 8-[(2R,3S)-3-octyloxiran-2-yl]octanoic acid |
| Molecular Weight (g/mol) | 298.46 |
| Show More |