missing translation for 'onlineSavingsMsg'
Learn More

rac 3-Hydroxybutyric Acid-13C4 Sodium Salt, TRC

Product Code. 30135362
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135362 5 mg 5mg
30135586 10 mg 10mg
30003425 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135362 Supplier Toronto Research Chemicals Supplier No. TRCH8330245MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 287111-43-7
Molecular Formula 13C4H7NaO3
Molecular Weight (g/mol) 130.06
Synonym 3-Hydroxybutanoic Acid-13C4 Sodium Salt, 3-Hydroxybutyric-13C4 Acid Monosodium Salt, DL-β-Hydroxybutyric-13C4 Acid Sodium Salt, Sodium 3-Hydroxybutyrate-13C4, Sodium Dl-β-hydroxybutyrate-13C4, Sodium β-Hydroxybutyrate-13C4, β-Hydroxybutyric-13C4 Acid Sodium Salt
IUPAC Name sodium 3-hydroxybutanoate-1,2,3,4-13C4
SMILES [13CH3][13CH](O)[13CH2][13C](O[Na])=O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C4H8O3.Na/c1-3(5)2-4(6)7;/h3,5H,2H2,1H3,(H,6,7);/q;+1/p-1/i1+1,2+1,3+1,4+1;
Formula Weight 130.04
Chemical Name or Material rac 3-Hydroxybutyric Acid-13C4 Sodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.