CAS RN 287111-43-7
CAS RN 287111-43-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
rac 3-Hydroxybutyric Acid-13C4 Sodium Salt, TRC
CAS: 287111-43-7 Molecular Formula: 13C4H7NaO3 Molecular Weight (g/mol): 130.06 Synonym: 3-Hydroxybutanoic Acid-13C4 Sodium Salt,3-Hydroxybutyric-13C4 Acid Monosodium Salt,DL-β-Hydroxybutyric-13C4 Acid Sodium Salt,Sodium 3-Hydroxybutyrate-13C4,Sodium Dl-β-hydroxybutyrate-13C4,Sodium β-Hydroxybutyrate-13C4,β-Hydroxybutyric-13C4 Acid Sodium Salt IUPAC Name: sodium 3-hydroxybutanoate-1,2,3,4-13C4 SMILES: [13CH3][13CH](O)[13CH2][13C](O[Na])=O