Learn More
Epredia™ RA Lamb Waxes and Embedding Media
Product Code. 12634077 Shop All Epredia Products
Change view
Click to view available options
Product Type:
Wax and Embedding Media
Quantity:
10 kg
1 x 10 kg
Unit Size:
10kg
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Quantity Unit Size
12634077 Wax and Embedding Media 1 x 10 kg Each
12624077 Wax and Embedding Media 10 kg 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12634077 Supplier Epredia™ Supplier No. W11/4
  • ONLINE EXCLUSIVE PRICE
    103.00 EUR
    79.66 EUR / Pack of 1
    Save 23.34 EUR
    22% Off
 Restricted Item
Estimated Shipment: 22-02-2027
Quickly and efficiently mount slides with Epredia™ RA Lamb waxing and embedding media for slide mounting.
Waxing and embedding media include:
  • Immersion oil
  • Embedding medium
  • Mounting medium
  • Chemical stain
  • Formaspill granules
  • Paraffin wax
  • Solution buffer
TRUSTED_SUSTAINABILITY
Specifications
Product Type Wax and Embedding Media
Quantity 1 x 10 kg
compliance-icons
Product Identifier
  • MEDIA RAYMOND A LAMB WAXING AND EMBEDDING MEDIA for slide mounting pure
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P391-Collect spillage.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Phenol