Learn More
(R)-tert-Butyl 3-(hydroxymethyl)piperidine-1-carboxylate, 97%, BLD Pharm™
Specifications
Specifications
| CAS | 140695-85-8 |
| Molecular Formula | C11H21NO3 |
| MDL Number | MFCD02683202 |
| Quantity | 5 g |
| SMILES | C(=O)(OC(C)(C)C)N1CCC[C@H](C1)CO |
| Molecular Weight (g/mol) | 215.29 |
| Percent Purity | 97% |
| Chemical Name or Material | (R)-tert-Butyl 3-(hydroxymethyl)piperidine-1-carboxylate |