Learn More
(R,S)-N-Nitroso Anatabine-2,4,5,6-d4 (40 ug/ml in Acetonitrile), TRC
Product Code. 30406134
Change view
Click to view available options
Quantity:
1x10 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30406134 1x10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 30406134 Lieferant Toronto Research Chemicals Lieferanten-Nr. TRCKIT41581X10ML
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Spezifikation
Chemical Name or Material (R,S)-N-Nitroso Anatabine-2,4,5,6-d4
CAS 1020719-69-0
Molecular Formula C10 2H4 H7 N3 O
Quantity 1x10 mL
Synonym 2,3'-Bipyridine-2',4',5',6'-d4, 1,2,3,6-tetrahydro-1-nitroso-, (RS)-N-Nitrosoanatabine D4 (pyridinyl D4), (R,S)-N-Nitroso Anatabine-2,4,5,6-d4, (R,S)-N-Nitrosoanatabine D4 (pyridinyl D4), 1,2,3,6-Tetrahydro-1-nitroso-2,3'-bipyridine-2',4',5',6'-d4 (ACI)
InChI Formula InChI=1S/C10H11N3O/c14-12-13-7-2-1-5-10(13)9-4-3-6-11-8-9/h1-4,6,8,10H,5,7H2/i3D,4D,6D,8D
SMILES [2H]c1nc([2H])c(C2CC=CCN2N=O)c([2H])c1[2H]
IUPAC Name 2,3,4,6-tetradeuterio-5-(1-nitroso-3,6-dihydro-2H-pyridin-2-yl)pyridine
Molecular Weight (g/mol) 193.24
Formula Weight 193.1153
Concentration or Composition (by Analyte or Components) 40 μg/mL
Mehr anzeigen Weniger anzeigen